Merck
  • Total synthesis of (-)-cinatrin C1 based on an In(OTf)3-catalyzed Conia-Ene reaction.
Total synthesis of (-)-cinatrin C1 based on an In(OTf)3-catalyzed Conia-Ene reaction.
The Journal of organic chemistry (2013-04-13)
Fumiya Urabe, Shunsuke Nagashima, Keisuke Takahashi, Jun Ishihara, Susumi Hatakeyama
ABSTRACT
The stereocontrolled total synthesis of (-)-cinatrin C1, a phospholipase A2 inhibitor, has been accomplished. The key feature includes the stereoselective construction of the highly substituted tetrahydrofuran core by In(OTf)3-catalyzed Conia-ene reaction of the oxygen-tethered acetylenic malonic ester followed by dihydroxylation with concomitant lactonization.
Sigma-Aldrich
Silver trifluoromethanesulfonate, ≥99%
Sigma-Aldrich
Silver trifluoromethanesulfonate, purum, ≥98.0% (Ag)
Sigma-Aldrich
Aluminum trifluoromethanesulfonate, 99.9% trace metals basis
Sigma-Aldrich
Silver trifluoromethanesulfonate, ≥99.95% trace metals basis
Sigma-Aldrich
Trifluoromethanesulfonic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Trifluoromethanesulfonic acid, reagent grade, 98%
Sigma-Aldrich
Zinc trifluoromethanesulfonate, 98%
Sigma-Aldrich
Lanthanum(III) trifluoromethanesulfonate, 99.999% trace metals basis