Merck
  • Isolation and characterization of a phenol-degrading strain of Alcaligenes sp. AM4.
Isolation and characterization of a phenol-degrading strain of Alcaligenes sp. AM4.
Acta biologica Hungarica (2013-04-10)
Amani M D Elahwany, Mona E M Mabrouk
ABSTRACT
Six isolates with phenol degrading ability were obtained from marine sediments by enrichment procedures and an isolate, AM4, was identified as Alcaligenes sp. by 16S rDNA sequencing. The Plackett-Burman design was applied to estimate the significance of culture medium components and conditions for phenol degradation by Alcaligenes sp. AM4. The resulting medium formula which was predicted to be near optimal was: phenol conc. (240 μg/ml), culture volume (37.5 ml), inoculum's size (0.15 ml), NH4SO4 (0.5 g/l), K2HPO4 (0.75 g/l), KH2PO4 (0.75 g/l), MgSO4 (0.3 g/l) and NaCl (0.25 g/l). Scanning electron microscopy was applied to cells exposed to phenol, and a larger cell size was detected, resulting in a reduced cell surface. This relative reduction of the cell surface represents a cellular mechanism to reduce the toxic effect of this environmental stress factor.
Product Description
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)