Merck
  • Purification of porcine pancreatic lipase by aqueous two-phase systems of polyethylene glycol and potassium phosphate.
Purification of porcine pancreatic lipase by aqueous two-phase systems of polyethylene glycol and potassium phosphate.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2013-04-09)
Yu-Jie Zhou, Cheng-Li Hu, Na Wang, Wei-Wei Zhang, Xiao-Qi Yu
ABSTRACT
An aqueous two-phase system (ATPS) was applied for the purification of porcine pancreatic lipase (PPL) from crude PPL using polyethylene glycol (PEG) and potassium phosphate. Phase diagrams for polyethylene glycol (PEG) and potassium phosphate dibasic were determined at room temperature to find an operating region to first form the ATPS. The PPL was preferentially partitioned into the PEG-rich phase in systems with molecular weights of 1000 and 1500 and concentrated in the phosphate-rich phase in systems with PEG of 4000. Moreover, instead of tie line length (TLL), we used a stability ratio without NaCl in the system, and we first applied fluorescence spectroscopy for the protein conformational analysis of the ATPS. The molecular weight of the purified lipase was determined to be approximately 52 kDa by SDS-PAGE. The enzyme was efficiently purified in PEG 1500/potassium phosphate (17/13, %) at a pH of 7.0 at 4 °C. This system obtained an enzyme partition coefficient of 12.7, an extraction efficiency of 94.7% and a purification factor of approximately 4. These results demonstrate that the aqueous two-phase system is a highly efficient method for PPL purification.
Sigma-Aldrich
Potassium phosphate tribasic, reagent grade, ≥98%
Sigma-Aldrich
Potassium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Potassium phosphate dibasic, reagent grade, ≥98.0%
Sigma-Aldrich
Potassium phosphate dibasic solution, 1.0 M
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate dibasic, meets USP testing specifications
Sigma-Aldrich
Potassium Phosphate Monobasic solution, reagent grade, 1.0 M in solution
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate dibasic, anhydrous, for luminescence, for molecular biology, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Potassium phosphate dibasic, ACS reagent, ≥98%
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%