Merck
Atypical pneumonia associated with a Mycoplasma isolate in a kitten.
The Canadian veterinary journal = La revue veterinaire canadienne (2013-04-02)
Yannick Bongrand, Marie-Claude Blais, Kate Alexander
ABSTRACT
An atypical case of Mycoplasma pneumonia with an unusual radiographic and computed tomographic pattern was diagnosed in a Siamese kitten. The cat showed no response to broad-spectrum antibiotic therapy including enrofloxacin. The administration of doxycycline led to a dramatic clinical and radiographic improvement.
Sigma-Aldrich
Doxycycline hyclate
Supelco
Doxycycline hyclate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Doxycycline hyclate, VETRANAL®, analytical standard
Sigma-Aldrich
Doxycycline monohydrate
Sigma-Aldrich
Doxycycline Hydrochloride, Ready Made Solution, 100 mg/mL in DMSO, ≥95.0% (HPLC)