Merck
  • Treatment of a simulated phenolic effluent by heterogeneous catalytic ozonation using Pt/Al2O3.
Treatment of a simulated phenolic effluent by heterogeneous catalytic ozonation using Pt/Al2O3.
Environmental technology (2013-03-28)
Ana S Fajardo, Rui C Martins, Rosa M Quinta-Ferreira
ABSTRACT
Non-catalytic and catalytic ozonation over Pt/Al2O3 were considered in the treatment of a synthetic effluent composed of six phenolic acids usually present in olive mill wastewaters. In both processes the medium pH affected the rate of ozone decomposition and the formation of hydroxyl radicals. The optimum values were achieved for the catalytic system under pH 7 with 93.0 and 47.7%, respectively, of total phenol content and chemical oxygen demand (COD) removal, after 120 minutes of reaction. For pH 3, the catalytic ozonation followed a free radical pathway perceived by the presence of radical scavengers. No significant structural differences were observed between the fresh and used solid catalyst in X-ray diffraction analysis. Aluminium leaching behaviour was also evaluated at the end of each experiment. Moreover, a sequence of feed-batch trials involving the catalyst reutilization exhibited almost constant activity during the operation time. Eco-toxicological tests were performed for both processes, revealing that the treated effluent still presents some ecological impact, although it is lower than that for the raw wastewater.
Sigma-Aldrich
Platinum, wire, diam. 0.127 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum black, fuel cell grade, ≥99.9% trace metals basis
Sigma-Aldrich
Platinum, gauze, 100 mesh, 99.9% trace metals basis
Sigma-Aldrich
Platinum black, low bulk density, ≥99.9% trace metals basis
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Sigma-Aldrich
Platinum, wire, diam. 0.076 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.025 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 2.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum black, black, powder, ≤20 μm, ≥99.95% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.127 mm, 99.99% trace metals basis
Supelco
Aluminum oxide, activated, neutral, Brockmann Activity I
Sigma-Aldrich
Platinum, shot, ≤3 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.25 mm, thermocouple grade
Sigma-Aldrich
Platinum, foil, thickness 0.025 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 1.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Sigma-Aldrich
Platinum, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.05 mm, 99.99% trace metals basis
Supelco
Aluminum oxide, for the determination of hydrocarbons
Sigma-Aldrich
Aluminum oxide, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Platinum, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Platinum, powder, 99.995% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.20 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.05 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Platinum, powder (coarse), 99.99% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanopowder, <50 nm particle size (TEM)