Merck
  • Inhibition of the renin-angiotensin system reduces the rise in serum aldosterone in acute coronary syndrome patients with preserved left ventricular function: observations from the AVANT GARDE-TIMI 43 trial.
Inhibition of the renin-angiotensin system reduces the rise in serum aldosterone in acute coronary syndrome patients with preserved left ventricular function: observations from the AVANT GARDE-TIMI 43 trial.
Clinical chemistry (2013-03-20)
Jacob A Udell, David A Morrow, Eugene Braunwald, Karl Swedberg, Christoph Bode, Nader Rifai, Patrick C Brunel, Margaret F Prescott, Fang Ren, Elaine B Hoffman, Benjamin M Scirica
ABSTRACT
Acute coronary syndrome (ACS) activates neurohormonal pathways, including elevations in circulating aldosterone, with deleterious cardiovascular effects. We aimed to determine if early, more complete renin-angiotensin-aldosterone system inhibition (RAASI) in post-ACS patients without ventricular dysfunction or heart failure would result in a graded reduction in aldosterone concentrations. We performed serial measurement of serum aldosterone within the Aliskiren and Valsartan to Reduce NT-proBNP via Renin-Angiotensin-Aldosterone-System Blockade (AVANT GARDE)-Thrombolysis in Myocardial Infarction (TIMI) 43 trial, a randomized double-blind, placebo controlled trial of RAASI by valsartan, aliskiren, or both in post-ACS patients with preserved ventricular function but increased natriuretic peptides. Aldosterone was measured at randomization and week 8. Median aldosterone concentrations were comparable across treatment arms at baseline (9.26 ng/dL; interquartile range 7.12-12.76; n = 1073). In the placebo group, there was a significant increase in aldosterone over 8 weeks (19.7% rise, 2.20 (0.36) ng/dL, P < 0.0001) that was significantly reduced across active RAASI therapies (1.36 (0.39) ng/dL with aliskiren; 1.02 (0.37) ng/dL with valsartan; and 0.85 (0.37) ng/dL with combination therapy, P trend = 0.008). Compared to placebo, RAASI monotherapy resulted in a pooled relative absolute aldosterone change of -1.01 (0.45) ng/dL (P = 0.026 vs placebo), and combination therapy resulted in a relative absolute aldosterone change of -1.35 (0.52) ng/dL (P = 0.01 vs placebo). No significant difference in aldosterone concentrations was achieved between dual vs single RAASI (P = 0.47). In ACS patients with preserved ventricular function but increased natriuretic peptides, serum aldosterone rises over time and is blunted by more complete RAASI. The clinical implications and role for RAASI in this population warrant further investigation.
Product Description
Valsartan, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Valsartan, ≥98% (HPLC)
Sigma-Aldrich
Aldosterone, ≥95% (HPLC)
Supelco
Aldosterone solution, 100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Valsartan for system suitability, European Pharmacopoeia (EP) Reference Standard