Merck
  • Synthesis and chirality control of bulk crystals and nanocrystals: from a right-handed nonpolar chain to a left-handed polar chain.
Synthesis and chirality control of bulk crystals and nanocrystals: from a right-handed nonpolar chain to a left-handed polar chain.
Inorganic chemistry (2013-03-20)
Yan-Yan Yin, Yan-Li Zhao, Jian-Gong Ma, Xiao-Chang Cao, Peng Cheng
ABSTRACT
Both bulk crystals and nanocrystals of two helical complexes, [Cu(μ2-L)(H2O)]n (1) and {[Cu(μ2-L)(H2O)]·2H2O}n (2) (H2L = thiazolidine-2,4-dicarboxylic acid), have been synthesized with the chiralities of right-handedness (1) and left-handedness (2), respectively. 4-Cyanopyridine and poly(vinylpyrrolidone) (PVP) have been applied to control the synthesis of complexes with different helicities in bulk-crystal and nanocrystal forms, respectively. 2 can be irreversibly transformed to 1 under heating. Associated with the conformation changing, the symmetry alters between nonpolar and polar space groups.
Sigma-Aldrich
Povidone, meets USP testing specifications
Sigma-Aldrich
Kollidon® 25
Supelco
Povidone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Polyvinylpyrrolidone, average Mw ~1,300,000 by LS
Sigma-Aldrich
Polyvinylpyrrolidone, K 90
Sigma-Aldrich
Polyvinylpyrrolidone solution, K 60, 45% in H2O
Sigma-Aldrich
Polyvinylpyrrolidone, K 30
Sigma-Aldrich
Polyvinylpyrrolidone, powder, average Mw ~29,000
Sigma-Aldrich
Polyvinylpyrrolidone, powder, average Mw ~55,000
Sigma-Aldrich
Polyvinylpyrrolidone, mol wt (number average molecular weight Mn 360kDa)
Sigma-Aldrich
Polyvinylpyrrolidone, nucleic acid hybridization tested, mol wt 360,000
Sigma-Aldrich
Polyvinylpyrrolidone, average mol wt 40,000
Sigma-Aldrich
Polyvinylpyrrolidone, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Polyvinylpyrrolidone, suitable for plant cell culture, average mol wt 10,000
Sigma-Aldrich
Polyvinylpyrrolidone, average mol wt 10,000
Sigma-Aldrich
Polyvinylpyrrolidone, K 25, tested according to Ph. Eur.