Merck
Otitis media.
Pediatric clinics of North America (2013-03-14)
Michael E Pichichero
ABSTRACT
Acute otitis media (AOM) is diagnosed based on visualization of a full or bulging tympanic membrane with middle ear effusion. The distribution of bacteria causing AOM in North America under the influence of pneumococcal conjugate vaccination and antibiotic selection pressure has resulted in a predominance of β-lactamase-producing Haemophilus influenzae followed by penicillin-resistant Streptococcus pneumoniae. Although guidelines continue to endorse amoxicillin as the preferred treatment, amoxicillin/clavulanate in high dosage would be the preferred treatment based on the otopathogen mix currently. Antibiotic prophylaxis has fallen into disfavor as a preventative strategy for AOM recurrences.
Product Description
Cefpodoxime proxetil, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Amoxicillin, 95.0-102.0% anhydrous basis
Supelco
Amoxicillin trihydrate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cefuroxime, VETRANAL®, analytical standard
Supelco
Amoxicillin trihydrate, VETRANAL®, analytical standard
Cefpodoxime proxetil for peak identification, European Pharmacopoeia (EP) Reference Standard
Cefpodoxime proxetil for impurity H identification, European Pharmacopoeia (EP) Reference Standard