Merck
  • Sulfonylation of quinoline N-oxides with aryl sulfonyl chlorides via copper-catalyzed C-H bonds activation.
Sulfonylation of quinoline N-oxides with aryl sulfonyl chlorides via copper-catalyzed C-H bonds activation.
Organic letters (2013-03-07)
Zhiyong Wu, Hongyu Song, Xiuling Cui, Chao Pi, Weiwei Du, Yangjie Wu
ABSTRACT
An efficient and concise one-pot protocol to synthesize sulfonylated quinoline N-oxides via copper-catalyzed C-H bond activation has been developed. Commercially available and less expensive aryl sulfonyl chlorides were used as the sulfonylation reagents. Various 2-aryl sulfonyl quinolines were obtained in up to 91% yields in chemo- and regioselective manners.
Sigma-Aldrich
Sulfuryl chloride, 97%