Merck
  • Discrimination of redox-responsible biomolecules by a single molecular sensor.
Discrimination of redox-responsible biomolecules by a single molecular sensor.
Organic letters (2013-03-07)
Jinrok Oh, Jong-In Hong
ABSTRACT
A new application of a fluorescent sensor (PyDPA) for the discrimination of redox-responsible molecules is reported. Nicotinamide adenine dinucleotide/nicotinamide adenine dinucleotide phosphate (NAD(+)/NADP(+)) and flavin mononucleotide/flavin adenine dinucleotide (FMN/FAD) were differentiated by means of ratiometric fluorescence change from excimer-monomer equilibrium and time-dependent fluorescence change, respectively.
Sigma-Aldrich
Riboflavin Sodium Phosphate Hydrate, tested according to Ph. Eur.
Sigma-Aldrich
Riboflavin 5′-monophosphate sodium salt hydrate, synthetic, ≥70% (HPLC)
Sigma-Aldrich
Riboflavin 5′-monophosphate sodium salt hydrate, 73-79% (fluorimetric)
Sigma-Aldrich
Riboflavin 5′-monophosphate sodium salt hydrate, suitable for electrophoresis, suitable for acrylamide photopolymerization, ≥70%, powder