Merck
  • Hyperthyroidism causes cardiac dysfunction by mitochondrial impairment and energy depletion.
Hyperthyroidism causes cardiac dysfunction by mitochondrial impairment and energy depletion.
The Journal of endocrinology (2013-02-23)
Sangeeta Maity, Dipak Kar, Kakali De, Vivek Chander, Arun Bandyopadhyay
ABSTRACT
This study elucidates the role of metabolic remodeling in cardiac dysfunction induced by hyperthyroidism. Cardiac hypertrophy, structural remodeling, and expression of the genes associated with fatty acid metabolism were examined in rats treated with triiodothyronine (T3) alone (8 μg/100 g body weight (BW), i.p.) for 15 days or along with a peroxisome proliferator-activated receptor alpha agonist bezafibrate (Bzf; 30 μg/100 g BW, oral) and were found to improve in the Bzf co-treated condition. Ultrastructure of mitochondria was damaged in T3-treated rat heart, which was prevented by Bzf co-administration. Hyperthyroidism-induced oxidative stress, reduction in cytochrome c oxidase activity, and myocardial ATP concentration were also significantly checked by Bzf. Heart function studied at different time points during the course of T3 treatment shows an initial improvement and then a gradual but progressive decline with time, which is prevented by Bzf co-treatment. In summary, the results demonstrate that hyperthyroidism inflicts structural and functional damage to mitochondria, leading to energy depletion and cardiac dysfunction.
Sigma-Aldrich
Anti-Porin (Ab-5) (185-197) Rabbit pAb, liquid, Calbiochem®
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Peroxisome Proliferator Activated Receptor α antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.
Sigma-Aldrich
Bezafibrate, ≥98%, solid
Pricing and availability is not currently available.