Merck
  • Field-collected permethrin-resistant Aedes aegypti from central Thailand contain point mutations in the domain IIS6 of the sodium channel gene (KDR).
Field-collected permethrin-resistant Aedes aegypti from central Thailand contain point mutations in the domain IIS6 of the sodium channel gene (KDR).
The Southeast Asian journal of tropical medicine and public health (2013-02-19)
Raweewan Srisawat, Narumon Komalamisra, Chamnarn Apiwathnasorn, Pungasem Paeporn, Sittiruk Roytrakul, Yupha Rongsriyam, Yuki Eshita
ABSTRACT
One of the mechanisms responsible for pyrethroid resistance in mosquitoes is mutations in domain IIS6 of voltage-gated sodium channel gene (kdr). Aedes aegypti larvae were collected from the central provinces of Thailand (Bangkok, Prachin Buri and Ratchaburi) and colonized until they became adults. Partial fragment of kdr of permethrin-resistant mosquitoes were amplified by RT-PCR and sequenced. Among the four nucleotide mutations detected, two mutations resulted in two amino acid substitutions, S(TCC) 989 P(CCC) and V(GTA)1016 G(GGA). Among 94 permethrin-resistant mosquitoes, the SS genotype (SS/VV) was found to predominate (n = 74), followed by SR (SP/VG) (n = 15) and RR (PP/ GG) genotypes (n = 5), with the resistant allele frequency ranging from 0.03 to 0.17. As pyrethroid insecticides are currently being advocated for use in Thailand, investigations of pyrethroid resistance in other regions of the country are needed to prevent potential cross-resistance among different types of insecticides.
Product Description
Supelco
Permethrin, PESTANAL®, analytical standard