Merck
  • The antimicrobial activity of the three commercially available intense sweeteners against common periodontal pathogens: an in vitro study.
The antimicrobial activity of the three commercially available intense sweeteners against common periodontal pathogens: an in vitro study.
The journal of contemporary dental practice (2013-02-14)
G M Prashant, Ravi B Patil, Tejavathi Nagaraj, Vinit B Patel
ABSTRACT
To evaluate the in vitro antimicrobial activity of three commercially available intense sweeteners against two common periodontal pathogens Porphyromonas gingivalis and Aggregatibacter actinomycetemcomitans. Three commercially available intense sweeteners namely saccharin, aspartame and sucralose were obtained and powdered. Necessary concentrations of the sweeteners were prepared by mixing them with an inert solvent. The antimicrobial efficacy was assessed using agar well diffusion technique. Statistical analysis was done using one-way ANOVA followed by Tukey's post hoc test. p-value < 0.05 was considered statistically significant. All the three sweeteners showed significant antimicrobial activity against the periodontal pathogens tested. Sucralose containing sucralose showed maximum zone of inhibition, against Aggregatibacter actinomycetemcomitans. Saccharin and aspartame containing saccharin and aspartame respectively, showed maximum zone of inhibition, against Porphyromonas gingivalis. All the sweeteners used in this study have demonstrated significant antimicrobial activity. Therefore, these sweeteners could be recommended as an ideal alternative to sucrose. Dental caries and periodontal diseases are ubiquitous diseases of mankind caused by microorganisms. Dental caries is caused by sucrose. By altering the source like intense sweetener we can combat caries as well as with its antimicrobial properties against periodontopathic bacteria, we can reduce prevalence of periodontal diseases.
Sigma-Aldrich
Saccharin, ≥98%
Sigma-Aldrich
Saccharin, ≥99%
Sigma-Aldrich
Sucralose, ≥98.0% (HPLC)
Supelco
Sucralose, analytical standard
Sigma-Aldrich
Asp-Phe methyl ester, ≥98%
Supelco
Mettler-Toledo Calibration substance ME 51143091, Saccharin, traceable to primary standards (LGC)
Supelco
Aspartame, analytical standard
Aspartame, European Pharmacopoeia (EP) Reference Standard