Merck
Illicit drugs: What dermatologists need to know.
Journal of the American Academy of Dermatology (2013-02-13)
Cara Hennings, Jami Miller
ABSTRACT
We review the most common systemic and cutaneous signs of heroin, cocaine, methamphetamine, Ecstasy, and marijuana use. We also provide an overview of the skin and soft-tissue infections frequently found in intravenous drug users and the effects of the adulterants added to the drugs.
Sigma-Aldrich
(−)-Tetramisole hydrochloride, ≥99% (GC)
Levamisole hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Levamisol hydrochloride, VETRANAL®, analytical standard
Levamisole hydrochloride for system suitability, European Pharmacopoeia (EP) Reference Standard