Merck
  • Successive alkali extraction and structural characterization of hemicelluloses from sweet sorghum stem.
Successive alkali extraction and structural characterization of hemicelluloses from sweet sorghum stem.
Carbohydrate polymers (2013-02-13)
Shao-Long Sun, Jia-Long Wen, Ming-Guo Ma, Run-Cang Sun
ABSTRACT
Sweet sorghum stem was successively extracted with water at 90 °C, 0.3, 0.6, 1.0, 1.5, and 2.0% KOH aqueous solution, and 60% ethanol containing 2.5% KOH at 75 °C for 3 h, yielding 76.3% of the original hemicelluloses. Chemical composition and structural characterization of the seven hemicellulosic fractions obtained were comparatively investigated by a combination of HPAEC, GPC, FT-IR, (1)H-, (13)C-, HSQC NMR and TGA techniques. According to the spectral analysis, hemicelluloses from sweet sorghum stem are assumed to L-arabino-4-O-methyl-D-glucurono-D-xylan. In addition, the higher molecular weights of hemicelluloses resulted in a higher thermal stability of the samples. The present study suggests that successive alkali extraction is a promising approach for fractionation of hemicelluloses from sweet sorghum stem and to prepare hemicellulosic polymers with different branching and molecular weights.
Sigma-Aldrich
Potassium hydroxide, tested according to Ph. Eur.
Sigma-Aldrich
Potassium hydroxide, ≥85% KOH basis, pellets, white
Sigma-Aldrich
Potassium hydroxide solution, 45 wt. % in H2O
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Sigma-Aldrich
Potassium hydroxide, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Potassium hydroxide, free-flowing, Redi-Dri, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Potassium hydroxide, pellets, reag. Ph. Eur., ≥85%
Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Potassium hydroxide, puriss. p.a., ≥86% (T), pellets
Sigma-Aldrich
Potassium hydroxide, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sigma-Aldrich
Potassium hydroxide, reagent grade, 90%, flakes
Sigma-Aldrich
Potassium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, 85-100.5%, pellets
Sigma-Aldrich
Potassium hydroxide, technical, ≥85%, powder
Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC