Merck
Challenges in detecting magnesium stearate distribution in tablets.
AAPS PharmSciTech (2013-02-05)
Satu Lakio, Balázs Vajna, István Farkas, Henri Salokangas, György Marosi, Jouko Yliruusi
ABSTRACT
Magnesium stearate (MS) is the most commonly used lubricant in pharmaceutical industry. During blending, MS particles form a thin layer on the surfaces of the excipient and drug particles prohibiting the bonding from forming between the particles. This hydrophobic layer decreases the tensile strength of tablets and prevents water from penetrating into the tablet restraining the disintegration and dissolution of the tablets. Although overlubrication of the powder mass during MS blending is a well-known problem, the lubricant distribution in tablets has traditionally been challenging to measure. There is currently no adequate analytical method to investigate this phenomenon. In this study, the distribution of MS in microcrystalline cellulose (MCC) tablets was investigated using three different blending scales. The crushing strength of the tablets was used as a secondary response, as its decrease is known to result from the overlubrication. In addition, coating of the MCC particles by MS in intact tablets was detected using Raman microscopic mapping. MS blending was more efficient in larger scales. Raman imaging was successfully applied to characterize MS distribution in MCC tablets despite low concentration of MS. The Raman method can provide highly valuable visual information about the proceeding of the MS blending process. However, the measuring set-up has to be carefully planned to establish reliable and reproducible results.
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG
Supelco
Stearic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Stearic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Stearic acid, analytical standard
Sigma-Aldrich
Stearic acid, reagent grade, 95%
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)
Supelco
Stearic Acid, Pharmaceutical Secondary Standard; Certified Reference Material