Merck
  • Prostaglandin E(2) mediates acid-induced heartburn in healthy volunteers.
Prostaglandin E(2) mediates acid-induced heartburn in healthy volunteers.
American journal of physiology. Gastrointestinal and liver physiology (2013-02-02)
Takashi Kondo, Tadayuki Oshima, Toshihiko Tomita, Hirokazu Fukui, Jiro Watari, Hiroki Okada, Shojiro Kikuchi, Mitsuru Sasako, Takayuki Matsumoto, Charles H Knowles, Hiroto Miwa
ABSTRACT
Prostaglandin E(2) (PGE(2)) plays a major role in pain processing and hypersensitivity. This study investigated whether PGE(2) levels are increased in the esophageal mucosa after acid infusion and whether increases in PGE(2) are associated with heartburn. Furthermore, expression of the PGE(2) receptor EP1 was investigated in human esophageal mucosa. Fourteen healthy male volunteers were randomized to 30-min lower esophageal acid (1% HCl) or saline perfusion. Before and after acid perfusion, endoscopic biopsies were taken from the distal esophagus. PGE(2) concentration (pg/mg protein) and EP1 mRNA and protein in biopsy samples were measured by ELISA, RT-PCR, and Western blotting. Symptom status of heartburn was evaluated with a validated categorical rating scale with a higher values corresponding to increasing intensity. PGE(2) levels in the esophageal mucosa significantly increased after acid infusion (before vs. after acid infusion: 23.2 ± 8.6 vs. 68.6 ± 18.3, P < 0.05), but not after saline infusion (before vs. after saline infusion: 9.3 ± 2.5 vs. 9.0 ± 3.2, NS). Time to first sensation (min) after acid infusion was less than after saline (saline vs. acid infusion: 22.1 ± 4.1 vs. 5.4 ± 1.5, P < 0.05). Intensity of heartburn in the acid-infusion group was also significantly greater compared with saline (saline vs. acid infusion: 54.3 ± 13.1 vs. 178.5 ± 22.8, P < 0.01). Changes in PGE(2) levels in the esophagus correlated with symptom intensity score (r = 0.80, P = 0.029). EP1 mRNA and protein expression were observed in the normal human esophageal mucosa. Esophageal PGE(2) expression is associated with mucosal acid exposure and heartburn.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Pricing and availability is not currently available.
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Pricing and availability is not currently available.
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Pricing and availability is not currently available.
Supelco
Hydrogen chloride - 1-butanol solution, ~3 M in 1-butanol, for GC derivatization, LiChropur
Pricing and availability is not currently available.