Merck
  • Update of dalbavancin spectrum and potency in the USA: report from the SENTRY Antimicrobial Surveillance Program (2011).
Update of dalbavancin spectrum and potency in the USA: report from the SENTRY Antimicrobial Surveillance Program (2011).
Diagnostic microbiology and infectious disease (2013-01-30)
Ronald N Jones, Helio S Sader, Robert K Flamm
ABSTRACT
Dalbavancin (DAL) is an investigational lipoglycopeptide with a prolonged serum half-life allowing once weekly dosing. DAL potency was assessed in the 2011 SENTRY Antimicrobial Surveillance Program among 1555 isolates sampled from all 9 US Census regions. Monitored Gram-positive cocci included Staphylococcus aureus (SA; 1,036/50.4% MRSA), coagulase-negative staphylococci (CoNS; 115), Enterococcus faecalis (25), E. faecium (31), Streptococcus pyogenes (155), Streptococcus agalactiae (153), and viridans group streptococci (VGS; 40). All susceptibility (S) testing used Clinical and Laboratory Standards Institute reference broth microdilution methods and interpretations. DAL (MIC(50/90), 0.06/0.06 μg/mL) was 8- and 16-fold more active than daptomycin (DAP) and vancomycin (VAN), respectively against SA, with MSSA and MRSA having the same MIC(90) results. CoNS was slightly more DAL-S (MIC(50), ≤0.03μg/mL). The highest staphylococcal DAL MIC was only 0.25 μg/mL. β-Haemolytic streptococci (βHS) and VGS had DAL MIC results ranging from ≤0.03 to 0.25 μg/mL (MIC(90), 0.06-0.12 μg/mL), and only enterococci showed elevated DAL MIC results. VanA phenotype-resistant E. faecalis or E. faecium had DAL MIC values at ≥1 μg/mL; VanB strains were DAL-S (MIC, ≤0.25 μg/mL). All cited DAL quantitative values were consistent with earlier surveillance data (2006-2009), without evidence of MIC creep. In conclusion, year 2011 SENTRY Program data for DAL documents sustained potent activity against SA, CoNS, βHS, VGS, and VAN-S enterococci, which averaged 4- to 32-fold greater than VAN, DAP, or linezolid.
Sigma-Aldrich
Clindamycin hydrochloride, lincosamide antibiotic
Pricing and availability is not currently available.
Clindamycin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Supelco
Clindamycin hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
Teicoplanin
Pricing and availability is not currently available.