Merck
  • Preparation of 3D nanoporous copper-supported cuprous oxide for high-performance lithium ion battery anodes.
Preparation of 3D nanoporous copper-supported cuprous oxide for high-performance lithium ion battery anodes.
Nanoscale (2013-01-29)
Dequan Liu, Zhibo Yang, Peng Wang, Fei Li, Desheng Wang, Deyan He
ABSTRACT
Three-dimensional (3D) nanoporous architectures can provide efficient and rapid pathways for Li-ion and electron transport as well as short solid-state diffusion lengths in lithium ion batteries (LIBs). In this work, 3D nanoporous copper-supported cuprous oxide was successfully fabricated by low-cost selective etching of an electron-beam melted Cu(50)Al(50) alloy and subsequent in situ thermal oxidation. The architecture was used as an anode in lithium ion batteries. In the first cycle, the sample delivered an extremely high lithium storage capacity of about 2.35 mA h cm(-2). A high reversible capacity of 1.45 mA h cm(-2) was achieved after 120 cycles. This work develops a promising approach to building reliable 3D nanostructured electrodes for high-performance lithium ion batteries.
Sigma-Aldrich
Copper(I) oxide, powder, ≤7 μm, 97%
Sigma-Aldrich
Copper(I) oxide, ≥99.99% trace metals basis, anhydrous