Merck
  • Quantitative proteomic analysis of N-linked glycoproteins in human tear fluid.
Quantitative proteomic analysis of N-linked glycoproteins in human tear fluid.
Methods in molecular biology (Clifton, N.J.) (2013-01-09)
Lei Zhou, Roger W Beuerman
ABSTRACT
Human tear fluid is a complex biological fluid that is actually the extracellular fluid of the surface of the eye and can be considered a dilute protein solution. The quality of the tears are critical for vision as they provide the mirror smooth reflex for the surface of the eye and hence focusing of images on the retina. Changes in tear composition may reflect the health of the epithelial cells lining the ocular surface and they have been shown to be useful to discover biomarkers for eye diseases. Glycoproteins are potentially important biomarkers of disease and therapeutic targets and can also be found in human tears. In this chapter, we concentrated on technical details in quantitative proteomic analysis of N-linked glycoproteins in human tears by combining hydrazide chemistry enrichment of N-linked glycoproteins and iTRAQ technology.
Sigma-Aldrich
Hydrazine sulfate salt, ACS reagent, ≥99.0%
Sigma-Aldrich
Hydrazine dihydrochloride, ≥98%
Sigma-Aldrich
Hydrazine sulfate salt, puriss. p.a., ACS reagent, ≥99.0%
Sigma-Aldrich
Hydrazine monohydrate, N2H4 64-65 %, reagent grade, ≥97%
Sigma-Aldrich
Hydrazine hemisulfate salt, ≥98%
Sigma-Aldrich
Hydrazine solution, 1.0 M in ethanol
Sigma-Aldrich
Hydrazine solution, 1 M in acetonitrile