Merck
Optical detection of lithocholic acid with liquid crystal emulsions.
Langmuir : the ACS journal of surfaces and colloids (2012-12-21)
Tanmay Bera, Jiyu Fang
ABSTRACT
The concentration level of bile acids is a clinical biomarker for the diagnosis of intestinal diseases because individuals suffering from intestinal diseases have a sharply increased concentration of bile acids at micromolar levels. Here, we report the detection of lithocholic acid (LCA) in aqueous solution by using surfactant-stabilized 4-n-pentyl-4'-cyanobiphenyl (5CB) liquid crystal droplets as an optical probe. We find that the surfactant adsorbed at the 5CB/water interface can be replaced by LCA, triggering a radial-to-bipolar configuration transition of the 5CB in the droplets. By simply observing the LCA-triggered transition with a polarizing optical microscope, micromolar levels of LCA in aqueous solution can be detected. The detection limit and selectivity of surfactant-stabilized 5CB droplets for LCA depend on the chain length and headgroup of the surfactants used for stabilizing 5CB droplets.
Product Description
Lithocholic acid, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Lithocholic acid, ≥95%
Pricing and availability is not currently available.