Merck
Design of irreversible optical nanothermometers for thermal ablations.
Chemical communications (Cambridge, England) (2012-12-12)
Tiffany P Gustafson, Qian Cao, Steven T Wang, Mikhail Y Berezin
ABSTRACT
Nanothermometers composed from a gold nanorod core, temperature sensitive linker and fluorescent dye are reported. The nanothermometers have low fluorescence due to a self-quenching mechanism at temperatures below 50 °C and become highly fluorescence above 70 °C.
Sigma-Aldrich
7-(Diethylamino)coumarin-3-carboxylic acid, BioReagent, suitable for fluorescence, ≥98.0% (HPCE)