Merck
  • A new model of reversible obstructive jaundice using rapidly absorbable suture materials.
A new model of reversible obstructive jaundice using rapidly absorbable suture materials.
Clinical and investigative medicine. Medecine clinique et experimentale (2012-12-12)
Nurettin Kahramansoy, Hayri Erkol, Edip E Yilmaz, Mustafa Şit, Fahri Yilmaz, Mehmet Tosun, Cavit Çöl
ABSTRACT
Reversible obstructive jaundice models have some limiting features, including the need for a second anaesthesia, re-laparotomy and surgical intervention after common bile duct ligation. The present study investigates the feasibility of a new application that can eliminate these limitations. Rapidly absorbable suture materials were used for ligation; therefore, spontaneous biliary decompression was anticipated by the self release of these rapidly degrading materials. Common bile ducts in Wistar Albino rats were ligated with silk, polyglytone 6211, or irradiated polyglactine 910 (n=7 for each group). Rats were grouped according to both the suture materials and the experiments termination date: 5 days (sham, silk5, polyglytone5, polyglactine5) and 21 days (silk21, polyglytone21, polyglactine21) after the ligation. Biochemical and morphologic changes of liver were assessed. The group polyglactine21 showed significantly lower mean ALT, AST, GGT, total and direct bilirubin values when compared with the group polyglactine5 (p=0.004-0.037). Morphologic changes did not correlate with the biochemical amelioration. In the group polyglytone21, not only the biochemical but also the morphologic changes significantly ameliorated when compared with the group polyglytone5 (p=0.003-0.043). No procedure associated mortality was observed. Common bile duct ligation with polyglytone offers a new reversible model for prolonged obstructive jaundice which abolishes the need for relaparotomy and a second surgical intervention and significantly reduces mortality.
Sigma-Aldrich
Resomer® RG 503 H, Poly(D,L-lactide-co-glycolide), acid terminated, lactide:glycolide 50:50, Mw 24,000-38,000
Sigma-Aldrich
Resomer® RG 502 H, Poly(D,L-lactide-co-glycolide), acid terminated, viscosity 0.16-0.24 dL/g 
Sigma-Aldrich
Resomer® RG 502, Poly(D,L-Lactide-co-Glycolide), lactide:glycolide 50:50, ester terminated, Mw 7,000-17,000
Sigma-Aldrich
Resomer® RG 752 H, Poly(D,L-lactide-co-glycolide), acid terminated, lactide:glycolide 75:25, Mw 4,000-15,000
Sigma-Aldrich
Resomer® RG 653 H, Poly(D,L-lactide-co-glycolide), acid terminated, Mw 24,000-38,000
Sigma-Aldrich
Resomer® RG 756 S, Poly(D,L-lactide-co-glycolide), ester terminated, lactide:glycolide 75:25, Mw 76,000-115,000
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), ester terminated, Mw 50,000-75,000
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), lactide:glycolide (75:25), mol wt 66,000-107,000
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), lactide:glycolide 65:35, Mw 40,000-75,000
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), lactide:glycolide (50:50), mol wt 30,000-60,000
Sigma-Aldrich
Resomer® RG 504, Poly(D,L-lactide-co-glycolide), lactide:glycolide 50:50, ester terminated, Mw 38,000-54,000
Sigma-Aldrich
Resomer® RG 858 S, Poly(D,L-lactide-co-glycolide), ester terminated, lactide:glycolide 85:15, Mw 190,000-240,000
Sigma-Aldrich
Resomer® RG 503, Poly(D,L-lactide-co-glycolide), lactide:glycolide 50:50, ester terminated, Mw 24,000-38,000
Sigma-Aldrich
Resomer® RG 504 H, Poly(D,L-lactide-co-glycolide), acid terminated, lactide:glycolide 50:50, Mw 38,000-54,000
Sigma-Aldrich
Resomer® RG 505, Poly(D,L-lactide-co-glycolide), ester terminated, Mw 54,000-69,000