Merck
  • Testing a biofilter cover design to mitigate dairy effluent pond methane emissions.
Testing a biofilter cover design to mitigate dairy effluent pond methane emissions.
Environmental science & technology (2012-12-12)
Chris Pratt, Julie Deslippe, Kevin R Tate
ABSTRACT
Biofiltration, whereby CH(4) is oxidized by methanotrophic bacteria, is a potentially effective strategy for mitigating CH(4) emissions from anaerobic dairy effluent lagoons/ponds, which typically produce insufficient biogas for energy recovery. This study reports on the effectiveness of a biofilter cover design at oxidizing CH(4) produced by dairy effluent ponds. Three substrates, a volcanic pumice soil, a garden-waste compost, and a mixture of the two, were tested as media for the biofilters. All substrates were suspended as 5 cm covers overlying simulated dairy effluent ponds. Methane fluxes supplied to the filters were commensurate with emission rates from typical dairy effluent ponds. All substrates oxidized more than 95% of the CH(4) influx (13.9 g CH(4) m(-3) h(-1)) after two months and continued to display high oxidation rates for the remaining one month of the trial. The volcanic soil biofilters exhibited the highest oxidation rates (99% removal). When the influx CH(4) dose was doubled for a month, CH(4) removal rates remained >90% for all substrates (maximum = 98%, for the volcanic soil), suggesting that biofilters have a high capacity to respond to increases in CH(4) loads. Nitrous oxide emissions from the biofilters were negligible (maximum = 19.9 mg N(2)O m(-3) h(-1)) compared with CH(4) oxidation rates, particularly from the volcanic soil that had a much lower microbial-N (75 mg kg(-1)) content than the compost-based filters (>240 mg kg(-1)). The high and sustained CH(4) oxidation rates observed in this laboratory study indicate that a biofilter cover design is a potentially efficient method to mitigate CH(4) emissions from dairy effluent ponds. The design should now be tested under field conditions.