MilliporeSigma
  • Effects of orally administered moxaverine on ocular blood flow in healthy subjects.
Effects of orally administered moxaverine on ocular blood flow in healthy subjects.
Graefe's archive for clinical and experimental ophthalmology = Albrecht von Graefes Archiv fur klinische und experimentelle Ophthalmologie (2012-12-04)
Doreen Schmidl, Berthold Pemp, Michael Lasta, Agnes Boltz, Semira Kaya, Stefan Palkovits, Franz Prager, Leopold Schmetterer, Gerhard Garhofer
ABSTRACT
To investigate the effect of orally administered moxaverine (Kollateral forte®) on ocular blood flow in young healthy subjects. Sixteen healthy subjects (eight male/eight female) aged between 20 and 32 years were included in this placebo-controlled, double-masked, two-way crossover study. Volunteers received 900 mg moxaverine-hydrochloride administered orally in three equal doses or placebo identical in appearance on 2 study days. Outcome variables were measured at baseline and 5 h after first drug administration. Laser Doppler flowmetry was used to assess choroidal and optic nerve head blood flow. Blood velocities in the retrobulbar vessels were measured with color Doppler imaging. Neither moxaverine nor placebo changed mean arterial pressure or intraocular pressure. Neither moxaverine nor placebo had an effect on choroidal (moxaverine: by 9.5 ± 17.2 %, placebo 3.8 ± 18.8 %, p = 0.54 between groups) or optic nerve head blood flow (moxaverine: 4.8 ± 10.4 %, placebo: 1.8 ± 10.9 %, p = 0.52 between groups). Similarly, administration of moxaverine did not change blood flow velocities or calculated resistance index in the retrobulbar vessels compared to placebo. The data of the present study indicate that orally administered moxaverine does not increase ocular blood flow. This is in contrast to previous findings, where parenteral administration of moxaverine lead to a significant increase in choroidal blood flow and blood flow velocities in the retrobulbar vessels. The reason for these differing results is unclear, but may be related to the low bioavailability after oral administration.
Product Description
Supelco
Papaverine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Papaverine hydrochloride, powder