Merck
Hydrotreatment of bio-oil over Ni-based catalyst.
Bioresource technology (2012-11-10)
Xinghua Zhang, Tiejun Wang, Longlong Ma, Qi Zhang, Ting Jiang
ABSTRACT
Inexpensive non-sulfided Ni-based catalysts were evaluated for hydrotreatments using phenol as model compound. HZSM-5, a zeolite with different ratio of Si/Al and γ-Al(2)O(3) were impregnated with Ni(NO(3))(2) · 6H(2)O and calcined at 450 °C. Conversion rates and product distribution for treatment of phenol at 160-240 °C in the presence of catalysts with nickel loads of 6, 10, 14 and 17 wt.% were determined. Phenol conversion was highest (91.8%) at 240 °C in the presence of HZSM-5(Si/Al = 38) loaded with 10% Ni. When hydrotreatment was carried out with bio-oil obtained from pyrolysis of pine sawdust under the optimal conditions determined for phenol, the pH of bio-oil increased from 2.27 to 4.07, and the hydrogen content increased from 6.28 to 7.01 wt.%. The decrease in acidity is desirable for the use of upgraded bio-oil.
Sigma-Aldrich
Nickel(II) nitrate hexahydrate, puriss. p.a., ≥98.5% (KT)
Sigma-Aldrich
Nickel(II) nitrate hexahydrate, crystals
Sigma-Aldrich
Nickel(II) nitrate hexahydrate, purum p.a., crystallized, ≥97.0% (KT)
Sigma-Aldrich
Nickel(II) nitrate hexahydrate, 99.999% trace metals basis