Merck
  • Occurrence and sensory perception of Z-2-(β-d-glucopyranosyloxy)-3-phenylpropenoic acid in rooibos (Aspalathus linearis).
Occurrence and sensory perception of Z-2-(β-d-glucopyranosyloxy)-3-phenylpropenoic acid in rooibos (Aspalathus linearis).
Food chemistry (2012-11-06)
Elizabeth Joubert, Dalene de Beer, Christiaan J Malherbe, Nina Muller, Susan L Bonnet, Jan H van der Westhuizen, Daneel Ferreira
ABSTRACT
Z-2-(β-d-glucopyranosyloxy)-3-phenylpropenoic acid (PPAG), a compound postulated to contribute to the taste and mouthfeel of fermented rooibos tea (Aspalathus linearis), was isolated from unfermented rooibos plant material. Its structure was unequivocally confirmed by LC-MS, -MS(2), FT-IR and NMR of the underivatised natural product, and optical rotation measurements of the hydrolysed sugar moiety. A similar compound, postulated to be E-2-(β-d-glucopyranosyloxy)-3-phenylpropenoic acid, was also detected. Analysis of the leaves of a large number of rooibos plants (n=54), sampled at commercial plantations, showed that PPAG is not ubiquitously present in detectable quantities in the leaves of different plants. This leads to large variation in the fermented plant material, infusions and food-grade extracts. PPAG was shown to have a slightly bitter to astringent taste and a detection threshold of 0.4 mg/l in water.
Sigma-Aldrich
Phenylpropiolic acid, 99%
Pricing and availability is not currently available.