Merck
  • Antrodia camphorata ATCC 200183 sporulates asexually in submerged culture.
Antrodia camphorata ATCC 200183 sporulates asexually in submerged culture.
Applied microbiology and biotechnology (2012-10-30)
Yan Geng, Zhe He, Zhen-Ming Lu, Hong-Yu Xu, Guo-Hua Xu, Jin-Song Shi, Zheng-Hong Xu
ABSTRACT
Antrodia camphorata is a well-known Chinese medicinal mushroom that protects against diverse health-related conditions. Submerged fermentation of A. camphorata is an alternative choice for the effective production of bioactive metabolites, but the effects of nutrition and environment on mycelial morphology are largely unknown. In this study, we show that A. camphorata American Type Culture Collection 200183 can form arthrospores in the end of liquid fermentation. Different morphologies of A. camphorata in submerged culture were analyzed using scanning electron microscopy. The optimal carbon and nitrogen sources for sporulation were soluble starch and yeast extract. We found that a carbon-to-nitrogen ratio (C/N) of 40:1, MgSO4 (0.5 g/l), KH2PO4 (3.0 g/l), an initial pH 5.0, and an inoculum size of 1.5×10(5) spores/ml led to maximum production of arthroconidia. Our results will be useful in the regulation and optimization of A. camphorata cultures for efficient production of arthroconidia in submerged culture, which can be used as inocula in subsequent fermentation processes.
Sigma-Aldrich
Potassium phosphate tribasic, reagent grade, ≥98%
Sigma-Aldrich
Potassium phosphate dibasic, anhydrous, for luminescence, for molecular biology, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Potassium phosphate dibasic, meets USP testing specifications
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium Phosphate Monobasic solution, reagent grade, 1.0 M in solution
Sigma-Aldrich
Potassium phosphate dibasic, reagent grade, ≥98.0%
Sigma-Aldrich
Potassium phosphate dibasic solution, 1.0 M
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Potassium phosphate dibasic, ACS reagent, ≥98%