Merck
  • A novel class of selective acetylcholinesterase inhibitors: synthesis and evaluation of (E)-2-(benzo[d]thiazol-2-yl)-3-heteroarylacrylonitriles.
A novel class of selective acetylcholinesterase inhibitors: synthesis and evaluation of (E)-2-(benzo[d]thiazol-2-yl)-3-heteroarylacrylonitriles.
Molecules (Basel, Switzerland) (2012-10-23)
Pedro de la Torre, Luis Astudillo Saavedra, Julio Caballero, Jairo Quiroga, Jans H Alzate-Morales, Margarita Gutiérrez Cabrera, Jorge Trilleras
ABSTRACT
(E)-2-(benzo[d]thiazol-2-yl)-3-heteroarylacrylonitriles are described as a new class of selective inhibitors of acetylcholinesterase (AChE). The most potent compound in the series exhibited good AChE inhibitory activity (IC₅₀ = 64 µM). Compound 7f was found to be more selective than galanthamine in inhibiting AChE and it showed a moderate selectivity index. Kinetic studies on AChE indicated that a competitive type of inhibition pattern exist for these acrylonitrile derivates. Molecular docking models of the ligand-AChE complexes suggest that compound 7 g is located on the periphery of the AChE active site.
Product Description
Supelco
Acrylonitrile, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Acrylonitrile, ≥99%, contains 35-45 ppm monomethyl ether hydroquinone as inhibitor
Pricing and availability is not currently available.
Sigma-Aldrich
Acrylonitrile, ≥99%, contains 35-45 ppm monomethyl ether hydroquinone as inhibitor
Pricing and availability is not currently available.
Supelco
Acrylonitrile solution, certified reference material, 5000 μg/mL in methanol
Pricing and availability is not currently available.