Merck
  • Synergy versus potency in the defensive secretions from nymphs of two pentatomomorphan families (Hemiptera: Coreidae and Pentatomidae).
Synergy versus potency in the defensive secretions from nymphs of two pentatomomorphan families (Hemiptera: Coreidae and Pentatomidae).
Journal of chemical ecology (2012-10-20)
Dorit Eliyahu, Roxanne A Ceballos, Vahid Saeidi, Judith X Becerra
ABSTRACT
One characteristic of true bugs (Heteroptera) is the presence of dorsal abdominal glands in the immature nymphal stages. These glands usually produce defensive chemicals (allomones) that vary among taxa but are still similar in closely related groups. Knowledge of the chemistry and prevalence of allomones in different taxa may clarify the evolution of these chemical defensive strategies. Within the infraorder Pentatomomorpha, the known secretions of nymphs of Pentatomidae tend to contain the hydrocarbon, n-tridecane, a keto-aldehyde, and an (E)-2-alkenal as the most abundant components. In the Coreidae, the dorsal abdominal gland secretions of nymphs often contain little or no hydrocarbon, and the most abundant keto-aldehyde and (E)-2-alkenal are often of shorter chain-length than those of pentatomids. We hypothesized that the long chain compounds would be less potent than their shorter homologs, and that bugs that carry the former would benefit from a synergistic effect of n-tridecane. To test this hypothesis we used three different behavioral assays with ants. A predator-prey assay tested the deterrence of allomones toward predators; a vapor experiment tested the effectiveness of allomones in the gaseous phase toward predators; and application of allomones onto predators tested the effect of direct contact. The results substantiate the hypothesis of a synergistic effect between n-tridecane and longer chain keto-aldehyde and (E)-2-alkenal in deterring predators. The short chain keto-aldehyde 4-oxo-(E)-2-hexenal was highly effective on its own. Thus, it seems that different groups of the infraorder diverged in their strategies involving defensive chemicals. Implications of this divergence are discussed.
Product Description
Supelco
Tridecane, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Tridecane, ≥99%
Pricing and availability is not currently available.