Merck
  • Impact of process variables on the micromeritic and physicochemical properties of spray-dried microparticles--Part II. Physicochemical characterisation of spray-dried materials.
Impact of process variables on the micromeritic and physicochemical properties of spray-dried microparticles--Part II. Physicochemical characterisation of spray-dried materials.
The Journal of pharmacy and pharmacology (2012-10-13)
Krzysztof J Paluch, Lidia Tajber, Maria I Amaro, Owen I Corrigan, Anne Marie Healy
ABSTRACT
In this work we investigated the residual organic solvent content and physicochemical properties of spray-dried chlorothiazide sodium (CTZNa) and potassium (CTZK) salts. The powders were characterised by thermal, X-ray diffraction, infrared and dynamic vapour sorption (DVS) analyses. Solvent levels were investigated by Karl-Fischer titration and gas chromatography. Spray-drying from water, methanol (MeOH) and mixes of MeOH and butyl acetate (BA) resulted in amorphous microparticles. The glass transition temperatures of CTZNa and CTZK were ∼192 and ∼159°C, respectively. These materials retained their amorphous nature when stored at 25°C in dry conditions for at least 6 months with no chemical decomposition observed. DVS determined the critical relative humidity of recrystallisation of CTZNa and CTZK to be 57% RH and 58% RH, respectively. The inlet temperature dependant oxidation of MeOH to formaldehyde was observed; the formaldehyde was seen to deposit within the amorphous matrix of spray-dried product. Spray-drying in the open blowing mode coupled with secondary drying resulted in a three-fold reduction in residual BA (below pharmacopoeial permitted daily exposure limit) compared to spray-drying in the closed mode. Experiments showed that recirculation of recovered drying gas increases the risk of deposition of residual solvents in the spray-dried product.
Product Description
Supelco
Chlorothiazide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chlorothiazide, thiazide diuretic
Chlorothiazide, European Pharmacopoeia (EP) Reference Standard