Merck
  • Synthesis and reduction kinetics of sterically shielded pyrrolidine nitroxides.
Synthesis and reduction kinetics of sterically shielded pyrrolidine nitroxides.
Organic letters (2012-10-12)
Joseph T Paletta, Maren Pink, Bridget Foley, Suchada Rajca, Andrzej Rajca
ABSTRACT
A series of sterically shielded pyrrolidine nitroxides were synthesized, and their reduction by ascorbate (vitamin C) indicate that nitroxide 3, a tetraethyl derivative of 3-carboxy-PROXYL, is reduced at the slowest rate among known nitroxides, i.e., at a 60-fold slower rate than that for 3-carboxy-PROXYL.
Sigma-Aldrich
Pyrrolidine, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Pyrrolidine, ≥99.5%, purified by redistillation
Pricing and availability is not currently available.
Sigma-Aldrich
Pyrrolidine, FG
Pricing and availability is not currently available.
Sigma-Aldrich
Pyrrolidine, ≥99.0%
Pricing and availability is not currently available.