Merck
  • Aspergillus galactomannan antigen assay and invasive aspergillosis in pediatric cancer patients and hematopoietic stem cell transplant recipients.
Aspergillus galactomannan antigen assay and invasive aspergillosis in pediatric cancer patients and hematopoietic stem cell transplant recipients.
Pediatric blood & cancer (2012-10-09)
Soo-Han Choi, Eun-Suk Kang, Hong Eo, So-Young Yoo, Ji Hye Kim, Keon Hee Yoo, Ki Woong Sung, Hong Hoe Koo, Yae-Jean Kim
ABSTRACT
Invasive aspergillosis (IA) is a major cause of morbidity and mortality in immunocompromised children. We investigated the usefulness of an Aspergillus galactomannan (GM) antigen assay as a diagnostic tool for IA in pediatric cancer patients and hematopoietic cell transplantation (HCT) recipients. The GM antigen assay results were analyzed in 749 blood samples from 99 patients. A GM index (GMI) greater than or equal to 0.5 on at least two separate occasions was considered positive. A review of the clinical data was performed for subjects with proven or probable IA. Twenty-one of 23 patients with proven or probable IA had positive GM antigen test results (91.3% sensitivity, 95% CI 71.9-98.9; 81.7% specificity, 95% CI 69.6-90.5; P < 0.0001). The false-positive rate was 18.3%. Being younger than 3 years of age, having a solid tumor, and receiving HCT within 4 weeks of the test were statistically significant factors for causing false-positive results (P < 0.05). Among the 23 patients with IA (six proven, 17 probable), 16 (69.6%) had hematological malignancies, five (22.7%) had solid tumors, and two (8.7%) had primary immunodeficiency. Nineteen patients (82.6%) received HCT. The most common clinical site of IA was the lungs (91.3%), and consolidation was the most frequent finding in chest CT scans (36.8%). The mortality at 12 weeks was 43.5%. Having a positive GM assay at least twice is useful in diagnosing IA in pediatric patients with cancer and HCT recipients.
Sigma-Aldrich
D-Galacto-D-mannan from Ceratonia siliqua, ~95% (HPLC)
Pricing and availability is not currently available.