Merck
  • Atomic layer engineering of perovskite oxides for chemically sharp heterointerfaces.
Atomic layer engineering of perovskite oxides for chemically sharp heterointerfaces.
Advanced materials (Deerfield Beach, Fla.) (2012-10-05)
Woo Seok Choi, Christopher M Rouleau, Sung Seok A Seo, Zhenlin Luo, Hua Zhou, Timothy T Fister, Jeffrey A Eastman, Paul H Fuoss, Dillon D Fong, Jonathan Z Tischler, Gyula Eres, Matthew F Chisholm, Ho Nyung Lee
ABSTRACT
Atomic layer engineering enables fabrication of a chemically sharp oxide heterointerface. The interface formation and strain evolution during the initial growth of LaAlO(3) /SrTiO(3) heterostructures by pulsed laser deposition are investigated in search of a means for controlling the atomic-sharpness of the interface. This study shows that inserting a monolayer of LaAlO(3) grown at high oxygen pressure dramatically enhances interface abruptness.
Sigma-Aldrich
Strontium titanate, powder, 99%
Sigma-Aldrich
Strontium titanate, nanopowder, <100 nm particle size, 99% trace metals basis
Sigma-Aldrich
Calcium titanate, nanopowder, <100 nm particle size (BET), 99% trace metals basis
Sigma-Aldrich
Strontium titanate, single crystal substrate, <100>