Merck
  • Enantioselective route to ketones and lactones from exocyclic allylic alcohols via metal and enzyme catalysis.
Enantioselective route to ketones and lactones from exocyclic allylic alcohols via metal and enzyme catalysis.
Organic letters (2012-09-26)
Madeleine C Warner, Anuja Nagendiran, Krisztián Bogár, Jan-E Bäckvall
ABSTRACT
A general and efficient route for the synthesis of enantiomerically pure α-substituted ketones and the corresponding lactones has been developed. Ruthenium- and enzyme-catalyzed dynamic kinetic resolution (DKR) with a subsequent Cu-catalyzed α-allylic substitution are the key steps of the route. The α-substituted ketones were obtained in high yields and with excellent enantiomeric excess. The methodology was applied to the synthesis of a naturally occurring caprolactone, (R)-10-methyl-6-undecanolide, via a subsequent Baeyer-Villiger oxidation.
Sigma-Aldrich
Allyl alcohol, ≥99%
Sigma-Aldrich
Allyl alcohol, ≥98.5%