Merck
  • Evidence for the involvement of the serotonergic 5-HT(1A) receptors in the antidepressant-like effect caused by hesperidin in mice.
Evidence for the involvement of the serotonergic 5-HT(1A) receptors in the antidepressant-like effect caused by hesperidin in mice.
Progress in neuro-psychopharmacology & biological psychiatry (2012-09-22)
Leandro C Souza, Marcelo G de Gomes, André T R Goes, Lucian Del Fabbro, Carlos B Filho, Silvana P Boeira, Cristiano R Jesse
ABSTRACT
The present study investigated a possible antidepressant-like activity of hesperidin using two predictive tests for antidepressant effect in mice: the forced swimming test (FST) and the tail suspension test (TST). Results demonstrated that hesperidin (0.1, 0.3 and 1 mg/kg, intraperitoneal, i.p.) decreased the immobility time in the FST and TST without affecting the locomotor activity in the open field test. The antidepressant-like effect of hesperidin (0.3 mg/kg) on the TST was prevented by the pretreatment of mice with p-chlorophenylalanine methyl ester (pCPA; 100 mg/kg, i.p., an inhibitor of serotonin synthesis) and WAY100635 (0.1 mg/kg, subcutaneous, s.c., a selective 5-HT(1A) receptor antagonist). Pretreatment of mice with prazosin (1 mg/kg, i.p., an α(1)-adrenoceptor antagonist), yohimbine (1 mg/kg, i.p., an α(2)-adrenoceptor antagonist), propranolol (2 mg/kg, i.p., a β-adrenoceptor antagonist), AMPT (100 mg/kg, i.p., an inhibitor of tyrosine hydroxylase), SCH23390 (0.05 mg/kg, s.c., a dopamine D(1) receptor antagonist), sulpiride (50 mg/kg, i.p., a dopamine D(2) receptor antagonist), ketanserin (1mg/kg, i.p., a 5-HT(2A/2C) receptor antagonist) or MDL72222 (1 mg/kg, i.p., a 5-HT(3) receptor antagonist) did not block the antidepressant-like effect of hesperidin (0.3 mg/kg, i.p.) in the TST. Administration of hesperidin (0.01 mg/kg, i.p.) and fluoxetine (1 mg/kg), at subeffective doses, produced an antidepressant-like effect in the TST. The antidepressant-like effect caused by hesperidin in mice in the TST was dependent on an interaction with the serotonergic 5-HT(1A) receptors. Taken together, these results suggest that hesperidin possesses antidepressant-like property and may be of interest source for therapeutic agent for the treatment of depressive disorders.
Product Description
Supelco
Hesperidin, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Hesperidin, ≥80% (HPLC)
Pricing and availability is not currently available.