Merck
  • Determination of glutamate uptake by high performance liquid chromatography (HPLC) in preparations of retinal tissue.
Determination of glutamate uptake by high performance liquid chromatography (HPLC) in preparations of retinal tissue.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2012-09-22)
Edinaldo Rogério da Silva Moraes, Alan Barroso Araújo Grisolia, Karen Renata Matos Oliveira, Domingos Luiz Wanderley Picanço-Diniz, Maria Elena Crespo-López, Caio Maximino, Evander de Jesus Oliveira Batista, Anderson Manoel Herculano
ABSTRACT
The present study describes a simple and efficient method utilizing high performance liquid chromatography (HPLC) coupled to fluorescence detection for the determination of kinetic parameters of glutamate uptake in nervous tissue. Retinal tissue obtained from 7-day-old chicks was incubated with known concentrations of glutamate (50-2000 μM) for 10 min, and the levels of the o-phtaldehyde (OPA)-derivatized neurotransmitter in the incubation medium were measured. By assessing the difference between initial and final concentrations of glutamate in the medium, a saturable uptake mechanism was characterized (K(m)=8.2 and V(max)=9.8 nmol/mg protein/min). This measure was largely sodium- and temperature-dependent, strongly supporting that the mechanism for concentration decrements is indeed uptake by high-affinity transporters. Added to this, our results also demonstrated that zinc chloride (an inhibitor of glutamate/aspartate transporters) evoked a concentration-dependent decrease in glutamate uptake, demonstrating the specificity of our methodology. Overall, the present work characterizes an alternative methodology to evaluate glutamate uptake in nervous tissue using HPLC. This approach could be an important tool for studies associated to the characterization of minute alterations in glutamate transport related with central nervous system injury.
Sigma-Aldrich
Zinc chloride, AnhydroBeads, amorphous, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Zinc chloride, AnhydroBeads, amorphous, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Zinc chloride, anhydrous, powder, ≥99.995% trace metals basis
Sigma-Aldrich
Zinc chloride, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Zinc chloride, 99.999% trace metals basis
Sigma-Aldrich
Zinc chloride 0.1 M solution
Sigma-Aldrich
Zinc chloride solution, 1.9 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Zinc chloride, reagent grade, ≥98%
Sigma-Aldrich
Zinc chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98-100.5%
Sigma-Aldrich
Zinc chloride, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Zinc chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Zinc chloride solution, 0.5 M in THF
Sigma-Aldrich
Zinc chloride, ACS reagent, ≥97%
Sigma-Aldrich
L-Homoserine