Merck
  • CaCl(2) as a bifunctional reusable catalyst: diversity-oriented synthesis of 4H-pyran library under ultrasonic irradiation.
CaCl(2) as a bifunctional reusable catalyst: diversity-oriented synthesis of 4H-pyran library under ultrasonic irradiation.
Molecular diversity (2012-09-13)
Hamid Reza Safaei, Mohsen Shekouhy, Athar Shirinfeshan, Sudabeh Rahmanpur
ABSTRACT
CaCl(2) is applied as an efficient reusable and eco-friendly bifunctional catalyst for the one-pot three-component synthesis of 4H-pyrans under ultrasonic irradiation. A broad range of substrates including the aromatic and heteroaromatic aldehydes, indoline-2,3-dione (isatin) derivatives, acenaphthylene-1,2-dione (acenaphthenequinone) and 2, 2-dihydroxy-2H-indene-1,3-dione (ninhydrin) were condensed with carbonyl compounds possessing a reactive α-methylene group and alkylmalonates. All reactions are completed in short times, and the products are obtained in good to excellent yields. The catalyst could be recycled and reused several times without any loss of efficiency.
Sigma-Aldrich
Ninhydrin, suitable for amino acid detection
Sigma-Aldrich
Ninhydrin, ACS reagent
Sigma-Aldrich
Isatin, 97%
Sigma-Aldrich
Acenaphthenequinone