Merck
  • Nanoaggregates of micropurified lipopolysaccharide identified using dynamic light scattering, zeta potential measurement, and TLR4 signaling activity.
Nanoaggregates of micropurified lipopolysaccharide identified using dynamic light scattering, zeta potential measurement, and TLR4 signaling activity.
Analytical biochemistry (2012-09-11)
Eugenio Hardy, Tobias Kamphuis, Aleksandre Japaridze, Jan C Wilschut, Mathias Winterhalter
ABSTRACT
Nanoaggregates composed of selected glycoforms from Escherichia coli 055:B5 lipopolysaccharide (LPS) were prepared by combining sodium dodecyl sulfate (SDS)-polyacrylamide gel electrophoresis, zinc-imidazole reverse staining, zinc chelation after cutting gel slices, elution with either 0.5% triethylamine (TEA) or 0.4% to 0.5% surfactant (SDS or deoxycholate [DOC]) from extrusion-generated gel microparticles, and centrifugal diafiltration after appropriate surfactant dilution. Dynamic light scattering allows detecting these aggregates, giving a size distribution from 10 to 100nm in diameter. The formation of the aggregates prepared with selected DOC-eluted LPS glycoforms was notably improved over those prepared with TEA-eluted glycoforms. As the O-side chain length increased in the composition of the former aggregates, there was a gradual decrease in the electrophoretic mobility (from -1.2 to 0.0110(-8)m(2)/Vs), giving a calculated zeta potential from -15 to 0.1mV at pH6.8. These aggregates were further characterized for their abilities to elicit agonistic effects on human Toll-like receptor 4, as shown by in vitro activation of nuclear factor kappa light chain enhancer of activated B cells (NF-κB) in engineered HEK293 cells.
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine hydrochloride, ≥99.0% (AT)