Merck
  • Label-free impedimetric thrombin sensor based on poly(pyrrole-nitrilotriacetic acid)-aptamer film.
Label-free impedimetric thrombin sensor based on poly(pyrrole-nitrilotriacetic acid)-aptamer film.
Biosensors & bioelectronics (2012-09-11)
Hui Xu, Karine Gorgy, Chantal Gondran, Alan Le Goff, Nicolas Spinelli, Christian Lopez, Eric Defrancq, Serge Cosnier
ABSTRACT
A label-free and highly sensitive impedimetric aptasensor was developed based on electropolymerized film for the determination of thrombin. The first step is the electrogeneration of a poly(pyrrole-nitrilotriacetic acid) (poly(pyrrole-NTA)) film onto the surface of electrodes followed by complexation of Cu(2+) ions. Then, the histidine labeled thrombin aptamer was immobilized onto the electrode through coordination of the histidine groups on the NTA-Cu(2+) complex. The aptamer sensor was applied for the detection and quantification of thrombin via impedimetric detection without a labeling step. A linear quantification of thrombin was obtained in the range 4.7×10(-12)-5.0×10(-10) mol L(-1) with a sensitivity of 2838 Ω/log unit (R(2)=0.9984). The impedance modulus at 0.3 Hz as a function of thrombin concentration was used to elaborate a similar linear relationship from 4.7×10(-12) to 5×10(-10) mol L(-1). In addition, aptamer-poly(pyrrole-NTA) electrodes incubated for 40 min in aqueous solutions of bovine serum albumin (BSA), lysozyme and IgG (5×10(-7) mol L(-1)) did not exhibit non-specific adsorption of proteins. Moreover, it has been demonstrated that the selective sensor can be regenerated several times with a good reproducibility.
Sigma-Aldrich
Nitrilotriacetic acid, ACS reagent, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Nitrilotriacetic acid, Sigma Grade, ≥99%
Pricing and availability is not currently available.