Merck
  • Polyethylene as a nonvolatile solid cosolvent phase for catalyst separation and recovery.
Polyethylene as a nonvolatile solid cosolvent phase for catalyst separation and recovery.
Journal of the American Chemical Society (2012-08-29)
Yanfei Yang, Nilusha Priyadarshani, Tatyana Khamatnurova, Jakkrit Suriboot, David E Bergbreiter
ABSTRACT
The studies described here show that a relatively low molecular weight, narrow polydispersity polyethylene (PE) wax (Polywax) can serve as a nontoxic and nonvolatile alternative to alkane solvents in monophasic catalytic organic reactions where catalysts and products are separated under biphasic conditions. In this application, a polymer that is a solid at room temperature substitutes for a conventional alkane solvent at ca. 80 °C. In addition to the advantages of being a nonvolatile, nontoxic, reusable solvent, this hydrocarbon polymer solvent, like heptane, can sequester nonpolar soluble polymer-bound catalysts after a reaction and separate them from products. The extent of this separation and its generality were studied using polyisobutylene (PIB)- and poly(4-dodecylstyrene)-bound dyes and PE-bound Pd allylic substitution catalysts, PIB-bound Pd cross-coupling catalysts, and PE- and PIB-bound metathesis catalysts. Catalytic reactions were effected using single-phase reaction mixtures containing Polywax with toluene, THF, or THF/DMF at ca. 80 °C. These solutions either separate into two liquid phases on addition of a perturbing agent or separate as a solid/liquid mixture on cooling. The hydrocarbon polymer-bound dyes or catalysts either separate into the hot liquid Polywax phase or coprecipitate with Polywax and are subsequently isolated as a nonvolatile Polywax solid phase that contains the dye or the recyclable catalyst.
Sigma-Aldrich
Polyisobutylene, average Mw ~500,000, average Mn ~200,000 by GPC/MALLS, average Mv ~420,000
Pricing and availability is not currently available.
Sigma-Aldrich
Polyisobutylene, average Mw ~1,000,000, average Mn ~600,000 by GPC/MALLS, average Mv ~1,200,000
Pricing and availability is not currently available.