Merck
  • Trimetallic magnesium complexes bearing amine-bis(benzotriazole phenolate) derivatives as bifunctional catalysts for ring-opening polymerization and CO2/epoxide coupling.
Trimetallic magnesium complexes bearing amine-bis(benzotriazole phenolate) derivatives as bifunctional catalysts for ring-opening polymerization and CO2/epoxide coupling.
Chemical communications (Cambridge, England) (2012-08-23)
Chen-Yu Li, Chen-Ruei Wu, Yi-Chang Liu, Bao-Tsan Ko
ABSTRACT
New trimetallic magnesium complexes were synthesized for bifunctional catalysis. Complex catalysed ε-caprolactone polymerizations in not only a 'living' manner but also in an 'immortal' fashion. Additionally, tri-Mg was active for the coupling of CO(2) with cyclohexene oxide (CHO) on addition of n-Bu(4)NBr co-catalysts to give cis-cyclohexene carbonate under mild conditions.
Sigma-Aldrich
Cyclohexene, contains 100 ppm BHT as inhibitor, ≥99.0%
Sigma-Aldrich
Cyclohexene, inhibitor-free, ReagentPlus®, 99%
Supelco
Cyclohexene, analytical standard, ≥99.5% (GC)
Sigma-Aldrich
ε-Caprolactone, 97%