Merck
  • High-detectivity multilayer MoS(2) phototransistors with spectral response from ultraviolet to infrared.
High-detectivity multilayer MoS(2) phototransistors with spectral response from ultraviolet to infrared.
Advanced materials (Deerfield Beach, Fla.) (2012-08-21)
Woong Choi, Mi Yeon Cho, Aniruddha Konar, Jong Hak Lee, Gi-Beom Cha, Soon Cheol Hong, Sangsig Kim, Jeongyong Kim, Debdeep Jena, Jinsoo Joo, Sunkook Kim
ABSTRACT
Phototransistors based on multilayer MoS(2) crystals are demonstrated with a wider spectral response and higher photoresponsivity than single-layer MoS(2) phototransistors. Multilayer MoS(2) phototransistors further exhibit high room temperature mobilities (>70 cm(2) V(-1) s(-1) ), near-ideal subthreshold swings (~70 mV decade(-1) ), low operating gate biases (<5 V), and negligible shifts in the threshold voltages during illumination.
Sigma-Aldrich
Molybdenum(IV) sulfide, powder
Sigma-Aldrich
Molybdenum(IV) sulfide, powder, <2 μm, 98%