Merck
  • Full utilization of a mass spectrometer using on-demand sharing with multiple LC units.
Full utilization of a mass spectrometer using on-demand sharing with multiple LC units.
Journal of mass spectrometry : JMS (2012-08-18)
Shu Li, Qin Hao, John Gounarides, Y Karen Wang
ABSTRACT
The applicability of liquid chromatography-mass spectrometry (LC/MS) is often limited by throughput. The sharing of a mass spectrometer with multiple LCs significantly improves throughput; however, the reported systems have not been designed to fully utilize the MS duty cycle, and as a result to achieve maximum throughput. To fully utilize the mass spectrometer, the number of LC units that a MS will need to recruit is application dependent and could be significantly larger than the current commercial or published implementations. For the example of a single analyte, the number may approach the peak capacity to a first degree approximation. Here, the construction of a MS system that flexibly recruits any number of LC units demanded by the application is discussed, followed by the method to port a previously developed LC/MS method to the system to fully utilize a mass spectrometer. To demonstrate the performance and operation, a prototypical MS system of eight LC units was constructed. When 1-min chromatographic separations were performed in parallel on the eight LCs of the system, the average LC/MS analysis time per sample was 10.5 s when applied to the analysis of samples in 384-well plate format. This system has been successfully used to conduct large-volume biochemical assays with the analysis of a variety of molecular entities in support of drug discovery efforts. Allowing the recruitment of the number of LC units appropriate for a given application, this system has the potential to be a plug-and-play system to fully utilize a mass spectrometer.
Sigma-Aldrich
Dioleoylglycerol, ≥99%, mixture of 1,3- and 1,2-isomers, liquid
Hydrocortisone acetate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydrocortisone 21-acetate, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone 21-acetate, meets USP testing specifications