Merck
  • MWCNT/V2O5 core/shell sponge for high areal capacity and power density Li-ion cathodes.
MWCNT/V2O5 core/shell sponge for high areal capacity and power density Li-ion cathodes.
ACS nano (2012-08-09)
Xinyi Chen, Hongli Zhu, Yu-Chen Chen, Yuanyuan Shang, Anyuan Cao, Liangbing Hu, Gary W Rubloff
ABSTRACT
A multiwall carbon nanotube (MWCNT) sponge network, coated by ALD V(2)O(5), presents the key characteristics needed to serve as a high-performance cathode in Li-ion batteries, exploiting (1) the highly electron-conductive nature of MWCNT, (2) unprecedented uniformity of ALD thin film coatings, and (3) high surface area and porosity of the MWCNT sponge material for ion transport. The core/shell MWCNT/V(2)O(5) sponge delivers a stable high areal capacity of 816 μAh/cm(2) for 2 Li/V(2)O(5) (voltage range 4.0-2.1 V) at 1C rate (1.1 mA/cm(2)), 450 times that of a planar V(2)O(5) thin film cathode. At much higher current (50×), the areal capacity of 155 μAh/cm(2) provides a high power density of 21.7 mW/cm(2). The compressed sponge nanoarchitecture thus demonstrates exceptional robustness and energy-power characteristics for thin film cathode structures for electrochemical energy storage.
Sigma-Aldrich
Vanadium(V) oxide, 99.9% trace metals basis
Sigma-Aldrich
Vanadium(V) oxide, ≥98%
Sigma-Aldrich
Vanadium(V) oxide, ≥99.6% trace metals basis