Merck
  • Ergogenic effects of β-alanine and carnosine: proposed future research to quantify their efficacy.
Ergogenic effects of β-alanine and carnosine: proposed future research to quantify their efficacy.
Nutrients (2012-08-02)
John Caruso, Jessica Charles, Kayla Unruh, Rachel Giebel, Lexis Learmonth, William Potter
ABSTRACT
β-alanine is an amino acid that, when combined with histidine, forms the dipeptide carnosine within skeletal muscle. Carnosine and β-alanine each have multiple purposes within the human body; this review focuses on their roles as ergogenic aids to exercise performance and suggests how to best quantify the former's merits as a buffer. Carnosine normally makes a small contribution to a cell's total buffer capacity; yet β-alanine supplementation raises intracellular carnosine concentrations that in turn improve a muscle's ability to buffer protons. Numerous studies assessed the impact of oral β-alanine intake on muscle carnosine levels and exercise performance. β-alanine may best act as an ergogenic aid when metabolic acidosis is the primary factor for compromised exercise performance. Blood lactate kinetics, whereby the concentration of the metabolite is measured as it enters and leaves the vasculature over time, affords the best opportunity to assess the merits of β-alanine supplementation's ergogenic effect. Optimal β-alanine dosages have not been determined for persons of different ages, genders and nutritional/health conditions. Doses as high as 6.4 g day(-1), for ten weeks have been administered to healthy subjects. Paraesthesia is to date the only side effect from oral β-alanine ingestion. The severity and duration of paraesthesia episodes are dose-dependent. It may be unwise for persons with a history of paraesthesia to ingest β-alanine. As for any supplement, caution should be exercised with β-alanine supplementation.
Sigma-Aldrich
β-Alanine, 99%
Sigma-Aldrich
β-Alanine, BioReagent, suitable for cell culture, suitable for insect cell culture
Supelco
β-Alanine, analytical standard
Sigma-Aldrich
β-Alanine, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
β-Alanine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L-Carnosine, ~99%, crystalline
Supelco
Beta Alanine, Pharmaceutical Secondary Standard; Certified Reference Material