Merck
  • Performance in anxiety and spatial memory tests following bilateral vestibular loss in the rat and effects of anxiolytic and anxiogenic drugs.
Performance in anxiety and spatial memory tests following bilateral vestibular loss in the rat and effects of anxiolytic and anxiogenic drugs.
Behavioural brain research (2012-07-25)
Yiwen Zheng, Irene Cheung, Paul F Smith
ABSTRACT
Vestibular dysfunction in humans is associated with anxiety and cognitive disorders. However, various animal studies of the effects of vestibular loss have yielded conflicting results, from reduced anxiety to increased anxiety, depending on the particular model of vestibular dysfunction and the anxiety test used. In this study we revisited the question of whether rats with surgical bilateral vestibular deafferentation (BVD) exhibit changes in anxiety-related behaviour by testing them in the open field maze (OFM), elevated plus maze (EPM) and elevated T maze (ETM) in the presence of a non-sedating anxiolytic drug, buspirone, or an anxiogenic drug, FG-7142. We also tested the animals in a spatial T maze (STM) in order to evaluate their cognitive function under the same set of conditions. We found that BVD animals exhibited increased locomotor activity (P≤0.003), reduced supported and unsupported rearing (P≤0.02 and P≤0.000, respectively) and reduced thigmotaxis (P≤0.000) in the OFM, which for the most part the drugs did not modify. By contrast, there were no significant differences between BVD and sham control animals in the EPM and the BVD animals exhibited a marginally longer escape latency in the ETM (P≤0.03), with no change in avoidance latency. In the STM, the BVD animals demonstrated a large and significant decrease in accuracy compared to the sham control animals (P≤0.000), which was not affected by drug treatment. These results have replicated previous findings regarding increased locomotor activity, reduced rearing and thigmotaxis in the OFM, and impaired performance in the STM. However, they failed to replicate some previous results obtained using the EPM and ETM. Overall, they do not support the hypothesis that BVD animals exhibit increased anxiety-like behaviour and suggest that the cognitive deficits may be independent of the emotional effects of vestibular loss.
Sigma-Aldrich
β-Carboline-3-carboxylic acid N-methylamide