Merck
  • The dioxin/POPs legacy of pesticide production in Hamburg: part 2--waste deposits and remediation of Georgswerder landfill.
The dioxin/POPs legacy of pesticide production in Hamburg: part 2--waste deposits and remediation of Georgswerder landfill.
Environmental science and pollution research international (2012-07-11)
Rainer Götz, Volker Sokollek, Roland Weber
ABSTRACT
α-HCH, β-HCH, and γ-HCH (lindane) were listed as persistent organic pollutants in the Stockholm Convention. Therefore, they need to be globally addressed including the wastes remaining from historic use and production. While at most lindane production sites the unintentionally produced 85% HCH waste isomers have been deposited, at a former pesticide factory in Hamburg-Moorfleet HCH waste isomers have been recycled from 1953 to 1984 by thermal decomposition to chlorobenzenes and resulted in high polychlorinated dibenzo-p-dioxin/polychlorinated dibenzofuran (PCDD/PCDF)-contaminated residues. The management of the PCDD/PCDF-contaminated waste from the former pesticide factory in Hamburg has been assessed and quantified. Based on past accredited PCDD/PCDF measurements, the registered 3,700 tonnes of disposed thermal HCH decomposition residue contained 333 to 854 kg of PCDD/PCDF toxicity equivalent (I-TEQ) in 53-102 tonnes total sum of PCDD/PCDF. The wastes have been deposited together with other wastes in landfills in Hamburg and other parts of Germany. For the Georgswerder landfill (Hamburg), where approximately 50% of the PCDD/PCDF is disposed, current and previous situation and remediation activities are described. While PCDD/PCDF leaching from the landfill is controlled and incinerated, more water soluble organochlorines (vinyl chloride, cis-1,2-dichlorethene, chlorobenzenes) and benzene remain as a challenge for groundwater management. A comprehensive aftercare program has been established and will need to be operated by future generations including renewal of containment systems. Former lindane/HCH productions need-in addition to HCH deposits-to be assessed for possible recycling practice of HCH and related PCDD/PCDF-containing deposits. This could systematically be addressed within the Stockholm Convention implementation.
Product Description
Supelco
2,4,5-T, PESTANAL®