Merck
  • Structure-activity effects in peptide self-assembly and gelation - Dendritic versus linear architectures.
Structure-activity effects in peptide self-assembly and gelation - Dendritic versus linear architectures.
Chemical communications (Cambridge, England) (2012-07-05)
Cecile A Lagadec, David K Smith
ABSTRACT
We demonstrate that linear-dendritic shape-isomerism can have an impact on the gelation potential of peptides based on lysine, but the architectural effects can be inverted depending on the choice of functional groups, with the nature of these protecting groups dominating the gelation ability.
Sigma-Aldrich
1,2-Dichlorobenzene solution, NMR reference standard, 5% in acetone-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Pricing and availability is not currently available.
Supelco
1,2-Dichlorobenzene, PESTANAL®, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
1,2-Dichlorobenzene, anhydrous, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
1,2-Dichlorobenzene, suitable for HPLC, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
1,2-Dichlorobenzene, ReagentPlus®, 99%
Pricing and availability is not currently available.