Merck
  • Degradation of carbofuran and carbofuran-derivatives in presence of humic substances under basic conditions.
Degradation of carbofuran and carbofuran-derivatives in presence of humic substances under basic conditions.
Chemosphere (2012-06-22)
Jorge Morales, José A Manso, Antonio Cid, Juan C Mejuto
ABSTRACT
The influence of humic aggregates in water solution upon the chemical stability of carbofuran (CF) and the carbofuran-derivatives, 3-hydroxy-carbofuran (HCF) and 3-keto-carbofuran (KCF), has been investigated in basic media. An inhibition upon the basic hydrolysis of 3-hydroxy-carbofuran and 3-keto-carbofuran (≈ 1.7 and ≈ 1.5-fold, respectively) was observed and it was rationalized in terms of the micellar pseudophase model. Nevertheless, non-significant effect upon the carbofuran stability was found in the presence of humic substances. These behaviors have been compared with the corresponding ones in other synthetic colloidal aggregates.
Sigma-Aldrich
Carbofuran, 98%